ContactUsLinkComponent
Racks for Gilson Pipetman pipette tips
Racks for Gilson Pipetman pipette tips
Catalog # GREI973270_50
Supplier:  Greiner Bio-One
Racks for Gilson Pipetman pipette tips
Catalog # GREI973270_50
Supplier:  Greiner Bio-One
Specifications
  • Description:
    Empty PP rack for pipette tips (10 - 200 µl), suitable for Gilson®
  • Pk:
    50
Specifications
About this item
Empty racks for 10 - 200 µl pipette tips that are ideally suited for Gilson Pipetman, Biohit, Brand, Eppendorf, Gilson and Socorex pipettes.