Fluosorber filter cannister
Catalog # HARV34-0415
Supplier: HARVARD APPARATUS
Specifications
- Description:Fluosorber filter canister
- Pk:6
Specifications
About this item
Activated charcoal canister that is designed to optimise halogenated gas absorption. Fluosorber is factory tested with an air flow of 60 LPM to ensure resistance is not above 2 cm H₂O.
- Designed to scavenge 200 g of anesthetic agent
- Stand-alone for passive scavenging or used with the Fluovac for efficient active scavenging
- Weighs 1200 g before use and 1400 g when full