ContactUsLinkComponent
Particulate filter, P3 R
Particulate filter, P3 R
Catalog # BART922404
Supplier:  BARTELS RIEGER
undefined
Particulate filter, P3 R
Catalog # BART922404
Supplier:  BARTELS RIEGER
Specifications
  • Type:
    Particle filter
  • Pk:
    5
  • Class:
    P3
Specifications
About this item
The high-performance particle filter for filtering or purifying air has a high-quality housing made from plastic.
  • Particle separation efficiency of >99,95%
  • Low inhalation resistance
  • Low weight and compact design
The particle filter is designed for use in areas contaminated by dusts and aerosols.
Certifications: EN 133, DIN EN 148-1, EN 143:200