ContactUsLinkComponent
Beakers, tall form, with spout
Beakers, tall form, with spout
Catalog # 213-1140
Supplier #: 1229
Beakers, tall form, with spout
Catalog # 213-1140
Supplier #: 1229
Log in for availability
Select an Option
60 mm
Select an Option
250 ml
Select an Option
115 mm
Select an Option
02
Select an Option
1
About this item
Quartz glass.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.
Certifications: DIN 12330.
Frequently Bought Together