Bulk media for ion exchange chromatography columns, PL-SCX
Fournisseur: VARIAN
Certificats
À propos de cet article
PL-SCX is a macroporous PS/DVB matrix with a very hydrophilic coating and strong cation exchange functionality. The manufacturing process is controlled to provide optimum density of strong cation-exchange moieties for analysing, separating, and purifying a wide range of biomolecules from small peptides to large proteins.
- Reliable higher-resolution separations
- Exceptional stability for long column lifetime
- Chemical stability for the widest range of eluents and clean-up procedures for purification
Frequently Bought Together