Sliding shelf racks for cryoboxes, Comfort
Anbieter: TENAK
Zertifikate
Über diesen Artikel
Stainless steel.
- Racks for five different box heights are available
- Can be used with all freezer brands
- Fits all standard cryoboxes
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility. Measure the inside height and depth of your freezer in order to choose the right rack.
Ordering information: Delivered with or without cryoboxes.
Frequently Bought Together