ContactUsLinkComponent
Test kits, nitrite, Comparator 2000+
Test kits, nitrite, Comparator 2000+
Catalog # LOVI502371
Supplier:  Lovibond
CAS Number:  
undefined
Test kits, nitrite, Comparator 2000+
Catalog # LOVI502371
Supplier:  Lovibond
CAS Number:  
Specifications
  • Chemical method:
    N-(1-Naphthyl)ethylenediamine
  • Pk:
    250
  • Description:
    Nitrite acidifying tablet
  • Range:
    0,002 – 0,05 mg/l N
Specifications
About this item
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.