ContactUsLinkComponent
Reagent for nickel determination using Lovibond® MD photometers
Reagent for nickel determination using Lovibond® MD photometers
Catalog # LOVI2419033
Supplier:  Lovibond
CAS Number:  
undefined
Reagent for nickel determination using Lovibond® MD photometers
Catalog # LOVI2419033
Supplier:  Lovibond
CAS Number:  
Specifications
  • Chemical method:
    Dimethylglyoxime
  • Pk:
    50 Tests
  • Description:
    Nickel-51, Nickel-52 reagent test
  • Range:
    0.02 - 1 mg/l, 0.2 - 7 mg/l Ni