Order Entry
Switzerland
ContactUsLinkComponent
Fluoride standard, Lovibond
Fluoride standard, Lovibond
Catalog # LOVI205630
Supplier:  Lovibond
CAS Number:  
Fluoride standard, Lovibond
Catalog # LOVI205630
Supplier:  Lovibond
CAS Number:  

Specifications

  • Concentration:
    1 mg/l
  • Matrix:
    H₂O
  • Pk:
    30 mL
  • Description:
    Fluoride standard solution
  • Environmentally Preferable:

Specifications

About this item