TAPI-2
Catalog # ENZOBMLPI1350001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:TAPI-2
- Purity:>99% (HPLC)
Specifications
About this item
Inhibitor of TNF-alpha processing
Inhibitor of TACE, other ADAMs, ACE secretase and other metalloproteinases. Blocks release of TNFα, TGF-α, DCC, APP, L-selectin, ACE, IL6R, NOTCH, and erbB2/HER2. Has been used in culture (typical concentration 5 to 100 µM).