E-64-D
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Calpain and cathepsin inhibitor
Cell-permeable, ethyl ester of E-64-c. The ester is hydrolyzed by intracellular esterases to produce E-64-c. Inhibits calpain and cathepsins B, H, and L. Inhibits degradation of autophagic cargo inside autophagolysosomes.
Specifications
- Protein/peptide name:E-64-D
- Purity:>98% (TLC)