Human SOS-1
Catalog # ENZOBMLP3050001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Species:Human
- Seq:02
- Protein/peptide name:SOS-1
- Purity:>98%
Specifications
About this item
PI3 kinase inhibitor
SH3 binding sequence of guanine nucleotide exchange factor hSOS which is a ligand for the adaptor protein Grb2 SH3 domain. Blocks SOS/Grb2 interaction and thereby prevents Ras activation via receptor protein tyrosine kinases. Km=5,6 µM for N-terminal SH3 of Grb2.