Order Entry
Switzerland
ContactUsLinkComponent
PKI (14-22) amide (myristoylated)
PKI (14-22) amide (myristoylated)
Catalog # ENZOBMLP2100500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PKI (14-22) amide (myristoylated)
Catalog # ENZOBMLP2100500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Seq:
    02
  • Protein/peptide name:
    PKI (14-22) amide (myristoylated)
  • Purity:
    >95%

Specifications

About this item

PKA inhibitor

Heat-stable protein kinase inhibitor (PKI) peptide sequence (14-22). N-terminus is myristoylated to increase cell membrane permeability. The non-myristoylated version of this peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA) with Ki=36 nM. This product is a useful tool for studying PKA in cellular systems.