Order Entry
Switzerland
ContactUsLinkComponent
mLCK
mLCK
Catalog # ENZOBMLP2090001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
mLCK
Catalog # ENZOBMLP2090001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    mLCK
  • Purity:
    97%

Specifications

About this item

MLCK inhibitor

This sequence, KRRWKKNFIAV, is from skeletal muscle myosin light chain kinase (MLCK) (342-352) pseudosubstrate site. Inhibits MLCK (Ki =1 µM with MLC peptide substrate). Possesses only slight calmodulin binding activity.