ContactUsLinkComponent
CaM kinase II (290-309)
CaM kinase II (290-309)
  ENZOBMLP2000500
 :  
CaM kinase II (290-309)
  ENZOBMLP2000500
 :  
  • Pk:
    0,5 mg
  • Seq:
    02
  • Protein/peptide name:
    CaM kinase II (290-309)
  • Purity:
    97%
CaM kinase inhibitor
Synthetic peptide with sequence from Ca²⁺/calmodulin-dependent protein kinase II pseudosubstrate site. Peptide binds calmodulin tightly and inhibits calmodulin-requiring enzymes including CaMKII, phosphodiesterase and calcineurin. IC50=52 nM for CaMKII in the presence of 100 nM calmodulin.