GP antagonist-2A
Catalog # ENZOBMLG5150001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:GP antagonist-2A
- Formulation:Lyophilised
Specifications
About this item
G protein antagonist
A G protein antagonist which selectively inhibits the activation of Gq by M1 muscarinic cholinergic receptors (mAChR). Not effective in inhibiting M2 mAChR-promoted GTP hydrolysis by Gi. Inhibition is reversible and competitive with respect to receptor binding to G proteins. Also inhibits activation by cholecystokinin receptor in pancreatic acini at 10 µM.