Z-D(OMe)E(OMe)VD(OMe)-FMK
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Inhibitor of caspase-3 and -7
Irreversible, cell permeable inhibitor of caspase-3 and -7.
Specifications
- Protein/peptide name:Z-D(OMe)E(OMe)VD(OMe)-FMK
- Purity:>95% (TLC)
- Formulation:Lyophilized
Specifications
Frequently Bought Together