Ac-LEVD-CHO
Catalog # ENZOALX260065M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Ac-LEVD-CHO
- Purity:>95% (HPLC)
- Formulation:Lyophilized
Specifications
About this item
Inhibitor of caspase-4 and -5
Reversible inhibitor of caspase-4. Also inhibits caspase-5.