Order Entry
Switzerland
ContactUsLinkComponent
Streptomyces chromofuscus phospholipase D
Streptomyces chromofuscus phospholipase D
Catalog # ENZOBMLSE3010025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Streptomyces chromofuscus phospholipase D
Catalog # ENZOBMLSE3010025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 KU
  • Species:
    Streptomyces chromofuscus
  • Seq:
    02
  • Form:
    Liquid
  • Enzyme name:
    Phospholipase D
  • Enzyme activity:
    25 kU
  • Purity:
    ≥90% (SDS-PAGE)
  • Concentration:
    11,5 mg/ml
  • Formulation:
    In 100 mM TRIS/HCl, pH 8,0, containing 10% glycerol and 0,1% Triton X-100.

Specifications

About this item

Isolated from Streptomyces chromofuscus.