Human recombinant Caspase-4 (from E. coli)
Catalog # ENZOBMLSE1765000
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 Vial
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Environmentally Preferable:
- Enzyme name:Caspase-4
- Enzyme activity:5000 U
Specifications
About this item
Produced in E. coli.