Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Caspase-10 (from E. coli)
Human recombinant Caspase-10 (from E. coli)
Catalog # ENZOBMLSE1745000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Caspase-10 (from E. coli)
Catalog # ENZOBMLSE1745000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Caspase-10
  • Enzyme activity:
    5000 U
  • Concentration:
    100 U/µl. One U=1 pmol/min using Ac-IETD-pNA (200 µM; cat# P-431) as substrate at 30°C

Specifications

About this item

Produced in E. coli.