Human recombinant Casein kinase II (from E. coli)
Catalog # ENZOBMLSE1240010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 KU
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Form:Liquid
- Enzyme name:Casein kinase II
- Enzyme activity:10 kU
- Purity:Free from proteases and phosphatases.
- Concentration:~0,7 mg/ml
- Formulation:In 20 mM TRIS-HCl, pH 7,5, containing 0,35 M sodium chloride, 1 mM EDTA, 2 mM DTT and 0,1% Triton X-100.
Specifications
About this item
Produced in E. coli. Consists of 44 kDa alpha catalytic and 26 kDa & szlig; regulatory subunits as an alpha2& szlig;2 tetramer.