Pig 12-Lipoxygenase (from Leukocytes)
Catalog # ENZOBMLPL0120100
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 Vial
- Source:Leukocytes
- Species:Pig
- Seq:02
- Environmentally Preferable:
- Form:Liquid
- Enzyme name:12-Lipoxygenase
- Enzyme activity:100 U
- Purity:Semi-purified
- Formulation:In 20 mM Tris·HCl buffer, pH 7,2, containing 0,5 mM EDTA, PMSF and DTT
Specifications
About this item
Isolated from pig leukocytes.