Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Caspase-1 (from E. coli)
Human recombinant Caspase-1 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Caspase-1 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Form:
    Lyophilised
  • Reconstitution instructions:
    Reconstitute to 1 U/µl with PBS containing 15% glycerol.
  • Enzyme name:
    Caspase-1
  • Enzyme activity:
    25 U

Specifications

Frequently Bought Together