Mouse recombinant IL-7
ENZOENZPRT1380010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Protein synonyms:Interleukin-7
- Protein/peptide name:IL-7
- Purity:>98% (SDS-PAGE gel & HPLC)
- Molecular weight:~15 kDa
- Formulation:Lyophilized
Produced in E.coli.