Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant NGAL
Mouse recombinant NGAL
Catalog # ENZOENZPRT1141000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant NGAL
Catalog # ENZOENZPRT1141000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Seq:
    02
  • Protein synonyms:
    Neutrophil gelatinase-associated lipocalin|Siderocalin|Lipocalin 2|Oncogene 24p3
  • Protein/peptide name:
    NGAL
  • Concentration:
    1000 pg/ml
  • Formulation:
    Liquid, In 0,01 M phosphate buffer, pH 7,4, containing 0,14 M NaCl, 0,5% BSA, 0,5% Tween 20, 0,0016% Phenol red, and 0,1% Proclin 950

Specifications

About this item

Produced in E. coli.