Human recombinant TARC
20 µG
Catalog # ENZOALX2018160020
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Produced in E. coli. Non-glycosylated protein, containing 71 amino acids.
- High purity
- Carrier-free
Specifications
Specifications
- Pk:20 µG
- Cat. No.:ENZOALX2018160020
- Protein/peptide type:Recombinant
- Protein/peptide name:TARC
- Formulation:Lyophilized from 0,02% TFA. Sterile filtered.
- Protein synonyms:ABCD-2|SCYA17|Thymus and activation regulated chemokine|CCL17
- Purity:>98% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Species:Human
- Biological activity:The activity is determined by the ability to chemoattract human T cells which is typically observed at a concentration of 2 - 40 ng/ml.
- Molecular weight:~8,1kDa
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



