Human recombinant IGF-I (from E. coli)
Catalog # ENZOADI9080590100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Biological activity:The activity as determined by the dose-dependent proliferation of mouse FDC-P1 is less than 1.0 ng/ml, corresponding to a specific activity of 1x106 units/mg.
- Protein synonyms:Insulin-like growth factor I
- Protein/peptide name:IGF-I
- Purity:>98% (SDS-PAGE; RP-HPLC)
- Molecular weight:~8 kDa
- Formulation:Lyophilized. Sterile filtered freeze-dried powder after extensive dialysis against 50 mM acetic acid.
- Purification:Purified by multi-step chromatography.