Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human native factor Vlla (from plasma)

Human native factor Vlla (from plasma)

10 µG
Catalog # ENZOBMLSE4380010
Supplier:   Enzo Life Sciences
Human native factor Vlla (from plasma)
Catalog # ENZOBMLSE4380010
Supplier:  Enzo Life Sciences
CAS Number:  
Human native factor Vlla (from plasma)

10 µG

CAS Number:  
Price
SFr. 538.00
Log in for availability
1 * 10 µG
Product Details & Documents

Isolated from human plasma and activated.

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    ENZOBMLSE4380010
  • Protein/peptide type:
    Native
  • Protein/peptide name:
    Factor Vlla
  • Grade:
    purified
  • Species:
    Human
  • Source:
    Plasma

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.