RGD control peptide
Catalog # ENZOBMLP7010005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:RGD control Peptide
- Purity:97%
- Tested applications:Control peptide
Specifications
About this item
Negative control for RGD peptide
Inactive control peptide.