SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Proteins and Peptides
Spantide II
Spantide II
ENZOBMLP5030001
:
ENZO LIFE SCIENCES
:
Spantide II
ENZOBMLP5030001
:
ENZO LIFE SCIENCES
:
Pk:
1 mg
Seq:
02
Protein/peptide name:
Spantide II
Purity:
97%
Substance P antagonist
Potent substance P antagonist.