Order Entry
Switzerland
ContactUsLinkComponent
CdC2 Substrate (Biotin)
CdC2 Substrate (Biotin)
Catalog # ENZOBMLP2720001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
CdC2 Substrate (Biotin)
Catalog # ENZOBMLP2720001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Biotin
  • Seq:
    02
  • Protein/peptide name:
    CDc2 Substrate
  • Purity:
    >95% (HPLC)
  • Formulation:
    Lyophilized

Specifications

About this item

This peptide, (Ac-S)-PGRRRRK-biotin, is a substrate for CDC2 kinase.