HER2 Substrate,Biotin
ENZOBMLP2430001
:
- Pk:1 mg
- Conjugation:Biotin
- Seq:02
- Protein/peptide name:HER2 substrate
- Purity:95%
This peptide, Biotin-Ahx-GGMEDIYFEFMGGKKK, identified by screening a degenerate peptide library, is a substrate for several kinases, including HER-2/c-neu/erb-B-2, Tie-2, cFMS, and cKit.
The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.