Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Fyn kinase substrate

Fyn kinase substrate

1 mg
Catalog # ENZOBMLP2150001
Supplier:   Enzo Life Sciences
Fyn kinase substrate
Catalog # ENZOBMLP2150001
Supplier:  Enzo Life Sciences
CAS Number:  
Fyn kinase substrate

1 mg

CAS Number:  
Price
SFr. 168.00
Log in for availability
1 * 1 mg
Product Details & Documents

This peptide, EFGTYGTLSKKK, is derived from a peptide (pF4) discovered, through phage display, to be a very good substrate for Fyn kinase.

  • Its Km is estimated to be 70 µM

The addition of three lysines on the C-terminus allows this peptide to be used in phosphocellulose kinase assays.

Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    ENZOBMLP2150001
  • Protein/peptide name:
    Fyn kinase substrate
  • Purity:
    95%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.