PKCdelta Substrate
Catalog # ENZOBMLP1540001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:PKCdelta substrate
- Purity:>95% (HPLC)
Specifications
About this item
This peptide sequence, ARRKRKGSFFYGG, was selected from a peptide library for optimal phosphorylation by protein kinase C delta (PKCdelta).