H1-7
Catalog # ENZOBMLP1060001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:H1-7
- Purity:97%
Specifications
About this item
PKA substrate
This sequence, RRKASGP, is derived from calf thymus histone H1 containing Ser38. Peptide is phosphorylated by protein kinase A.