Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ac-IETD-AMC

Ac-IETD-AMC

1 mg
Catalog # ENZOALX260042M001
Supplier:   Enzo Life Sciences
Ac-IETD-AMC
Catalog # ENZOALX260042M001
Supplier:  Enzo Life Sciences
CAS Number:  
Ac-IETD-AMC

1 mg

CAS Number:  
Price
SFr. 89.60
Log in for availability
1 * 1 mg
Product Details & Documents

Caspase and granzyme substrate

Fluorogenic substrate for caspase-6 and -8 and granzyme B. Also a substrate for caspase-10.

Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    ENZOALX260042M001
  • Protein/peptide name:
    Ac-IETD-AMC
  • Purity:
    >98% (HPLC)

Specifications