Recombinant GroEL (from E. coli)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein/peptide name:GroEL
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, in 50 mM TRIS-hydrogen chloride, pH 7,5, containing 100 mM sodium chloride, 10 mM magnesium chloride, 5 mM DTT, and 0,05% sodium azide
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together