Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protein Linking and Labeling Systems
Green 496 dUTP (lyophilized)
Green 496 dUTP (lyophilized)
Green 496 dUTP (lyophilized)
Catalog # ENZOENZ42831L0050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Green 496 dUTP (lyophilized)
Catalog # ENZOENZ42831L0050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
50 Nmo
Description:
Green 496 dUTP (lyophilised)
Seq:
02
Purity:
≥93% (HPLC)
Formulation:
Lyophilised
Specifications
About this item
Product Details
Product Details