ContactUsLinkComponent
IHC PBS tablets
IHC PBS tablets
Catalog # ENZOADI9502400100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
IHC PBS tablets
Catalog # ENZOADI9502400100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Pk:
    100
  • Description:
    IHC PBS tablets
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
Solid. 100 ml PBS (1X) tablets containing 137 mM sodium chloride, 2.7 mM potassium chloride, and 10 mM phosphate buffer.