Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Caspase assay buffer

Caspase assay buffer

Caspase assay buffer, 20 mL
Codice catalogo ENZOBMLKI1110020
Nome del fornitore:   Enzo Life Sciences
Caspase assay buffer
Codice catalogo ENZOBMLKI1110020
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  
Caspase assay buffer

Caspase assay buffer, 20 mL

Numero CAS:  
Price
SFr. 61.60
Log in per disponibilità
1 * 20 mL
Product Details & Documents

50 mM HEPES, pH 7,4, 100 mM NaCl, 0,1% CHAPS, 10 mM DTT, 1 mM EDTA, 10% glycerol.

Specifiche

Specifiche

  • Pk:
    20 mL
  • Description:
    Caspase assay buffer

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.