Order Entry
Switzerland
ContactUsLinkComponent
BIO-PROBE® Lambda probe
BIO-PROBE® Lambda probe
Catalog # ENZOENZ40840
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
BIO-PROBE® Lambda probe
Catalog # ENZOENZ40840
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    2 µG
  • Application:
  • Description:
    BIO-PROBE® Lambda probe
  • Seq:
    02
  • Environmentally Preferable:
  • Size:
    2 µg

Specifications

About this item

Negative control probe for in situ hybridization.

The Lambda BIO-PROBE® labeled probe is prepared by nick translation of the lambda bacteriophage strain c1857ind1 Sam7. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).