ContactUsLinkComponent
AMPIGENE® HS Taq DNA Polymerase
AMPIGENE® HS Taq DNA Polymerase
Catalog # ENZOENZPRT1010500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
AMPIGENE® HS Taq DNA Polymerase
Catalog # ENZOENZPRT1010500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    AMPIGENE® HS Taq DNA polymerase
  • Seq:
    02
  • Units:
    500
Specifications
About this item
Increased sensitivity standard PCR for a broader range of samples, with enhanced speed, yield, and specificity.
AMPIGENE® HS Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE® HS Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. Hot-start technology is used to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine.The AMPIGENE® HS Taq DNA Polymerase is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine.
Ordering information: Contains AMPIGENE® HS Taq DNA polymerase (5u/µl) 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).