AMPIGENE® Taq mix
About this item
Optimised standard PCR mix for enhanced speed, yield, and specificity. Used for standard PCR applications.
AMPIGENE® Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq Mix is a robust mix for routine PCR applications including genotyping, screening, and library construction. An optimized buffer system allows efficient amplification under fast and standard cycling conditions.
: Kit contains 2×AMPIGENE® Taq Mix (AMPIGENE® Taq Polymerase, 6 mM MgCl₂, 2 mM dNTPs, enhancer and stabilizers).
3 Options Available Below
Product Details & Documents
Optimised standard PCR mix for enhanced speed, yield, and specificity. Used for standard PCR applications.
AMPIGENE® Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq Mix is a robust mix for routine PCR applications including genotyping, screening, and library construction. An optimized buffer system allows efficient amplification under fast and standard cycling conditions.
: Kit contains 2×AMPIGENE® Taq Mix (AMPIGENE® Taq Polymerase, 6 mM MgCl₂, 2 mM dNTPs, enhancer and stabilizers).
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ENZOENZNUC1000040
- ENZOENZNUC1000200
- ENZOENZNUC1001000
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- ENZOENZNUC1000040AMPIGENE® Taq mixSpecifications:
More
Specifications
Cat. No.ENZOENZNUC1000040Storage conditions−20 °C - ENZOENZNUC1000200AMPIGENE® Taq mixSpecifications:
More
Specifications
Cat. No.ENZOENZNUC1000200Storage conditions−20 °C - ENZOENZNUC1001000AMPIGENE® Taq mixSpecifications:
More
Specifications
Cat. No.ENZOENZNUC1001000Storage conditions−20 °C