Order Entry
Switzerland
ContactUsLinkComponent
Annexin V-EGFP apoptosis detection kit
Annexin V-EGFP apoptosis detection kit
Catalog # ENZOALX850253KI02
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Annexin V-EGFP apoptosis detection kit
Catalog # ENZOALX850253KI02
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Annexin V-EGFP apoptosis detection kit
  • Seq:
    02
  • Storage conditions:
    4 °C
  • Application:
    Flow Cytometry, Fluorescence Microscopy
  • Size:
    100 tests

Specifications

About this item

For the detection of apoptotic cells by flow cytometry.

EGFP (enhanced green fluorescent protein) has emerged as an ideal fluorochrome for FACS analysis and fluorescent microscopy as it offers a more intense signal than wild-type GFP. It has Ex(max): 488 nm and Em(max): 530 nm. The green signal produced makes EGFP ideal for dual staining with propidium iodide (PI).

Delivery: Kit contains Annexin V-EGFP, propidium iodide solution, binding buffer and assay protocol.