SOD activity kit
SOD Activity kit, 1 KIT
Catalog # ENZOADI900157
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
ELS offers a large collection of superoxide dismutase products for oxidative stress research.
For the measurement of total SOD activity in cell lysates and tissue from any species.
This is a versatile assay for the determination of SOD activity. Differentiate between the activity of three SOD isoenzymes: MnSOD, FeSOD, and Cu/ZnSOD. Obtain higher throughput sample analysis with a convenient colorimetric 96 well plate format and fast results in just 10 minutes.
Ordering information: Contents: Microtiter plate, Standard, SOD buffer concentrate, Xanthine oxidase, Xanthine solution, Triton x-100, WST-1 reagent
Specifications
Specifications
- Pk:1 KIT
- Description:SOD Activity kit
- Cat. No.:ENZOADI900157
- Size:96 wells