Human growth hormone ELISA kit
Codice catalogo ENZOENZKIT1480001
Nome del fornitore: ENZO LIFE SCIENCES
Numero CAS:
Specifiche
- Pk:1 KIT
- Primary antibody reactivity:Human
- Description:Human growth hormone ELISA kit
- Seq:01
- Sensitivity:0,93 pg/ml (3,91 - 250 pg/ml)
Specifiche
Informazioni su questo articolo
Ultra-sensitive human growth hormone ELISA kit enabling detection of HGH in serum, plasma, saliva and tissue culture media in just 2,5 hours.
- Ultra-sensitive measurement of HGH, detecting as little as 0,93 pg/ml
- Negligible cross reactivity with similar proteins (prolactin, human placental lactogen)
- High throughput format with results in 2,5 hours for up to 36 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of growth hormone in serum, plasma, saliva and tissue culture media. The human growth hormone ELISA kit is a colorimetric immunometric immunoassay kit with results in 2,5 hours.
Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.