Order Entry
Switzerland
ContactUsLinkComponent
Total PSA (human) ELISA kit
Total PSA (human) ELISA kit
Catalog # ENZOENZKIT1460001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Total PSA (human) ELISA kit
Catalog # ENZOENZKIT1460001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Description:
    Total PSA (Human) ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    0,287 pM or 0,024 ng/ml (333 - 1,37 pM or 28 - 0,12 ng/ml)

Specifications

About this item

Highly sensitive Total PSA ELISA kit enabling detection of Total PSA in human serum, plasma, urine, and tissue culture media samples in just 2,5 hours.

  • Highly-sensitive measurement of Total PSA, detecting as little as 0,287pM (or 0,024ng/ml) with negligible cross reactivity with similar proteins (KLK2, ACT)
  • Compatible with various sample types (plasma, serum, urine, tissue culture media)
  • High throughput format with results in 2,5 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of Total PSA in human serum, plasma, urine and tissue culture media. The Total PSA (human) ELISA kit is a colorimetric immunometric immunoassay kit with results in 2,5 hours.

Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.