KIM-1 (human) ELISA kit, grow cells
Katalognr. ENZOADI9002260001
Anbieter: ENZO LIFE SCIENCES
CAS-Nummer:
Technische Daten
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:KIM-1
- Description:KIM-1 (Human) ELISA kit, grow cells
- Seq:02
- Sensitivity:1,279 pg/ml (range 7,813 - 500 pg/ml)
Technische Daten
Über diesen Artikel
Ultra sensitive (1,2 pg/ml) ELISA kit, enabling reduced input sample and matrix interference, for the quantification of KIM-1, an early biomarker for kidney injury or disease.
- Sensitive: Reliably measure as little as 1,2 pg/ml of KIM-1
- High Throughput: Results in less than two hours from up to 38 samples in duplicate
- Specific: Low cross-reactivity to similar proteins
- Quantitative: Fully quantitative results for both normal and disease state samples that surpass semi-quantitative Western blot analysis
For the quantitative determination of human KIM-1 in urine.
The KIM-1 (human) ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of human KIM-1 in urine with results in less than two hours.
Ordering information: Contains Microtiter Plate, Assay Buffer, Standard, Antibody-conjugate, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate.