LBP, Soluble ELISA kit, Grow Cells
Catalog # ENZOALX850304KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:LBP, soluble ELISA kit, grow cells
- Seq:02
- Sensitivity:Range 5 - 50 ng/ml
Specifications
About this item
Quantitative determination of human, dog, pig, rabbit, bovine and horse-soluble LBP levels.
For the quantitative determination of human, dog, pig, rabbit, bovine and horse soluble LBP levels in serum, plasma and cell culture medium.
Ordering information: Contains 1 precoated ELISA module1 vial anti-LBP (human) detecting MAb1 vial LBP standard (human)1 vial reference serum2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution1 vial substrate solution.